accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOHO

CAS RN 57822-06-7

IUPAC Name: 5-oxononanedioic acid
Synonyms: 5-oxononanedioic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C9H14O5
InChi Key: GTCHZEFRDKAINX-UHFFFAOYSA-N
SMILES: OC(=O)CCCC(=O)CCCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results