accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONCH3OOCH3CH3H3C

CAS RN 57837-19-1

IUPAC Name: methyl 2-[N-(2,6-dimethylphenyl)-2-methoxyacetamido]propanoate
Synonyms: (+-)-metalaxyl subdue 5SP ridomil 2E (l-ala)-isomer ridomil caswell no. 375AA epa pesticide chemical code 113501 ridomil subdue ridomil, (L-ala)- metasyl
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: ZQEIXNIJLIKNTD-UHFFFAOYNA-N
SMILES: COCC(=O)N(C(C)C(=O)OC)C1=C(C)C=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results