accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5787-71-3

IUPAC Name: [(hydrazinecarbonyl)methyl]dimethylazanium
Synonyms: [(hydrazinecarbonyl)methyl]dimethylazanium
Molecular Weight (g/mol): 118.16
Molecular Formula: C4H12N3O
InChi Key: DALNXMAZDJRTPB-UHFFFAOYSA-O
SMILES: C[NH+](C)CC(=O)NN
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results