accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNHSN

CAS RN 5789-24-2

IUPAC Name: 5-[(1H-indol-3-yl)methyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 5-(1H-indol-3-ylmethyl)-3-phenyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 321.4
Molecular Formula: C18H15N3OS
InChi Key: KZXRWWVROVHSPI-UHFFFAOYNA-N
SMILES: O=C1C(CC2=CNC3=CC=CC=C23)NC(=S)N1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results