accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 579-07-7

IUPAC Name: 1-phenylpropane-1,2-dione
Synonyms: acetyl benzoyl acetylbenzoyl pyruvophenone phenylmethyldiketone benzoylacetyl
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: BVQVLAIMHVDZEL-UHFFFAOYSA-N
SMILES: CC(=O)C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)