accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57915-78-3

IUPAC Name: 1-(bromomethyl)-2,3-dichlorobenzene
Synonyms: 1-(bromomethyl)-2,3-dichlorobenzene
Molecular Weight (g/mol): 239.92
Molecular Formula: C7H5BrCl2
InChi Key: PVIGUZZDWGININ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(CBr)=C1Cl
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)