accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5794-88-7

IUPAC Name: 2-amino-5-bromobenzoic acid
Synonyms: 5-bromoanthranilic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: CUKXRHLWPSBCTI-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results