accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57966-95-7

IUPAC Name: (E)-2-[(ethylcarbamoyl)amino]-2-oxoethanecarbonimidoyl cyanide
Synonyms: cymoxanil
Molecular Weight (g/mol): 198.18
Molecular Formula: C7H10N4O3
InChi Key: XERJKGMBORTKEO-VZUCSPMQSA-N
SMILES: CCNC(=O)NC(=O)C(=N\OC)\C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results