accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57980-07-1

IUPAC Name: 4-methyl-7-(morpholin-4-yl)-2H-pyrano[2,3-b]pyridin-2-one
Synonyms: 4-methyl-7-(morpholin-4-yl)pyrano[2,3-b]pyridin-2-one
Molecular Weight (g/mol): 246.27
Molecular Formula: C13H14N2O3
InChi Key: UGHFUCYBDDPFJX-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=NC(=CC=C12)N1CCOCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results