accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58-39-9

IUPAC Name: 2-{4-[3-(2-chloro-10H-phenothiazin-10-yl)propyl]piperazin-1-yl}ethan-1-ol
Synonyms: perphenazine F-mon ethaperazine PZC 奋乃静 C21H26ClN3OS emesinal etaperazin decentan perphenazine preparation
Molecular Weight (g/mol): 403.97
Molecular Formula: C21H26ClN3OS
InChi Key: RGCVKNLCSQQDEP-UHFFFAOYSA-N
SMILES: OCCN1CCN(CCCN2C3=CC=CC=C3SC3=CC=C(Cl)C=C23)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results