accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONCH3NHNO

CAS RN 58-55-9

IUPAC Name: 1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: theophylline theotard pseudotheophylline xantivent Telb-DS dimethylxanthine solosin theoclair-SR medaphyllin theospan
Molecular Weight (g/mol): 180.17
Molecular Formula: C7H8N4O2
InChi Key: ZFXYFBGIUFBOJW-UHFFFAOYSA-N
SMILES: CN1C2=C(NC=N2)C(=O)N(C)C1=O
Molecular Weight (g/mol) 
  • (9)
  • (9)
Physical Form 
  • (6)
  • (6)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (6)
  • (3)
Grade 
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (6)
  • (2)
  • (4)
  • (6)
  • (3)
  • (5)
  • (6)
  • (9)
  • (9)
  • (6)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (6)
  • (3)
  • (1)
  • (6)