accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58-72-0

IUPAC Name: (2,2-diphenylethenyl)benzene
Synonyms: (2,2-diphenylethenyl)benzene benzilidenediphenylmethane
Molecular Weight (g/mol): 256.35
Molecular Formula: C20H16
InChi Key: MKYQPGPNVYRMHI-UHFFFAOYSA-N
SMILES: C(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)