accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3O

CAS RN 58-73-1

IUPAC Name: [2-(diphenylmethoxy)ethyl]dimethylamine
Synonyms: diphenhydramine aller-med diphen 苯海拉明 benzhydraminum medidryl diphenhydraminum diphenhydramine salicylate dibondrin hydramine
Molecular Weight (g/mol): 255.36
Molecular Formula: C17H21NO
InChi Key: ZZVUWRFHKOJYTH-UHFFFAOYSA-N
SMILES: CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results