accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClClCl

CAS RN 58-90-2

IUPAC Name: 2,3,4,6-tetrachlorophenol
Synonyms: chlorophenols
Molecular Weight (g/mol): 231.88
Molecular Formula: C6H2Cl4O
InChi Key: VGVRPFIJEJYOFN-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)