accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58-94-6

IUPAC Name: 6-chloro-1,1-dioxo-4H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
Synonyms: chlorothiazide chlorthiazidum sk-chlorothiazide chlorthiazide chlotride chloriazid diuresal 氯噻嗪 chlorthiazid alurene
Molecular Weight (g/mol): 295.71
Molecular Formula: C7H6ClN3O4S2
InChi Key: JBMKAUGHUNFTOL-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)