accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3O

CAS RN 580-02-9

IUPAC Name: methyl 2-(acetyloxy)benzoate
Synonyms: acetylsalicylic acid, methyl ester
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: ONWPLBKWMAUFGZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results