accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NN

CAS RN 580-15-4

IUPAC Name: quinolin-6-amine
Synonyms: 6-aminoquinoline
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: RJSRSRITMWVIQT-UHFFFAOYSA-N
SMILES: NC1=CC=C2N=CC=CC2=C1
Boiling Point 
  • (4)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (4)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (4)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (7)
  • (4)
  • (2)
  • (3)
  • (2)