accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NN

CAS RN 580-17-6

IUPAC Name: quinolin-3-amine
Synonyms: 3-aminoquinoline
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: SVNCRRZKBNSMIV-UHFFFAOYSA-N
SMILES: NC1=CN=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)