accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2N

CAS RN 5802-60-8

IUPAC Name: 1,1-dibenzylhydrazine
Synonyms: 1,1-dibenzylhydrazine
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: CZVXEAWVGZTLON-UHFFFAOYSA-N
SMILES: NN(CC1=CC=CC=C1)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results