accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 5802-82-4

IUPAC Name: methyl hexacosanoate
Synonyms: methyl hexacosanoate
Molecular Weight (g/mol): 410.73
Molecular Formula: C27H54O2
InChi Key: VHUJBYYFFWDLNM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)