accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58047-42-0

IUPAC Name: N-(4-bromophenyl)-4-methyl-N-(4-methylphenyl)aniline
Synonyms: N-(4-bromophenyl)-4-methyl-N-(4-methylphenyl)aniline
Molecular Weight (g/mol): 352.28
Molecular Formula: C20H18BrN
InChi Key: YMNJJMJHTXGFOR-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results