CAS RN 58066-85-6
IUPAC Name:
hexadecyl 2-(trimethylazaniumyl)ethyl phosphate
Synonyms:
miltex miltefosine HDPC miltefosin miltefosina miltefosinum baxter brand of miltefosine prasfarma brand of miltefosine baxter oncology brand of miltefosine choline phosphate hexadecyl ester, hydroxide, inner salt
Molecular Weight (g/mol):
407.58
Molecular Formula:
C21H46NO4P
InChi Key:
PQLXHQMOHUQAKB-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCOP([O-])(=O)OCC[N+](C)(C)C
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Miltefosine, 98%
Thermo Scientific Chemicals Miltefosine, 98%
CAS: 58066-85-6 Molecular Formula: C21H46NO4P Molecular Weight (g/mol): 407.57 InChI Key: PQLXHQMOHUQAKB-UHFFFAOYSA-N PubChem CID: 3599 ChEBI: CHEBI:75283