accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3OHOHOO

CAS RN 5809-23-4

IUPAC Name: 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoic acid
Synonyms: 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoic acid
Molecular Weight (g/mol): 313.35
Molecular Formula: C18H19NO4
InChi Key: FQNKTJPBXAZUGC-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C(=O)C2=CC=CC=C2C(O)=O)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results