CAS RN 581-40-8
2,3-Dimethylnaphthalene, 97%
CAS: 581-40-8 Molecular Formula: C12H12 Molecular Weight (g/mol): 156.228 MDL Number: MFCD00004119 InChI Key: WWGUMAYGTYQSGA-UHFFFAOYSA-N Synonym: guajen,naphthalene, 2,3-dimethyl,unii-35dj6se17o,2,3-dmn,2,3-dimethyl naphthalene,acmc-1ao3c,2,3-dimethylnaphthalene,naphthalene, 2,3-dimethyl-8ci 9ci PubChem CID: 11386 ChEBI: CHEBI:48615 IUPAC Name: 2,3-dimethylnaphthalene SMILES: CC1=CC2=CC=CC=C2C=C1C
2,3-Dimethylnaphthalene, 97%
CAS: 581-40-8 Molecular Formula: C12H12 Molecular Weight (g/mol): 156.23 InChI Key: WWGUMAYGTYQSGA-UHFFFAOYSA-N Synonym: guajen,naphthalene, 2,3-dimethyl,unii-35dj6se17o,2,3-dmn,2,3-dimethyl naphthalene,acmc-1ao3c,2,3-dimethylnaphthalene,naphthalene, 2,3-dimethyl-8ci 9ci PubChem CID: 11386 ChEBI: CHEBI:48615 IUPAC Name: 2,3-dimethylnaphthalene SMILES: CC1=CC2=CC=CC=C2C=C1C
2,3-Dimethylnaphthalene 98.0+%, TCI America™
CAS: 581-40-8 Molecular Formula: C12H12 Molecular Weight (g/mol): 156.228 MDL Number: MFCD00004119 InChI Key: WWGUMAYGTYQSGA-UHFFFAOYSA-N Synonym: guajen,naphthalene, 2,3-dimethyl,unii-35dj6se17o,2,3-dmn,2,3-dimethyl naphthalene,acmc-1ao3c,2,3-dimethylnaphthalene,naphthalene, 2,3-dimethyl-8ci 9ci PubChem CID: 11386 ChEBI: CHEBI:48615 IUPAC Name: 2,3-dimethylnaphthalene SMILES: CC1=CC2=CC=CC=C2C=C1C
Sigma Aldrich 2,3-Dimethylnaphthalene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
2,3-Dimethylnaphthalene, SPEX CertiPrep™
CAS: 581-40-8 Molecular Formula: C12H12 Molecular Weight (g/mol): 156.228 InChI Key: WWGUMAYGTYQSGA-UHFFFAOYSA-N PubChem CID: 11386 ChEBI: CHEBI:48615 IUPAC Name: 2,3-dimethylnaphthalene SMILES: CC1=CC2=CC=CC=C2C=C1C