accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOH

CAS RN 581-43-1

IUPAC Name: naphthalene-2,6-diol
Synonyms: 2,6-naphthalenediol
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: MNZMMCVIXORAQL-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=C(O)C=C2C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)