accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 581-45-3

IUPAC Name: 4-(piperidin-4-yl)pyridine
Synonyms: 4-(piperidin-4-yl)pyridine
Molecular Weight (g/mol): 162.24
Molecular Formula: C10H14N2
InChi Key: RGBWBVGQZFJTEO-UHFFFAOYSA-N
SMILES: C1CC(CCN1)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results