accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 581-46-4

NN

CAS RN 581-46-4

IUPAC Name: 3,3'-bipyridine
Synonyms: 3,3'-bipyridine
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: OFDVABAUFQJWEZ-UHFFFAOYSA-N
SMILES: C1=CN=CC(=C1)C1=CC=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results