accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5810-56-0

IUPAC Name: N-(piperidin-4-yl)acetamide
Synonyms: N-(piperidin-4-yl)acetamide
Molecular Weight (g/mol): 142.2
Molecular Formula: C7H14N2O
InChi Key: YLWUSMHZABTZGP-UHFFFAOYSA-N
SMILES: CC(=O)NC1CCNCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results