accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58111-94-7

IUPAC Name: 5-{[amino(nitroamino)methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 5-{[amino(nitroamino)methylidene]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 441.44
Molecular Formula: C21H23N5O6
InChi Key: RXMHIKWOZKQXCJ-UHFFFAOYNA-N
SMILES: NC(N[N+]([O-])=O)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)