accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5813-89-8

IUPAC Name: thiophene-2-carboxamide
Synonyms: thiophen-2-carboxamide
Molecular Weight (g/mol): 127.16
Molecular Formula: C5H5NOS
InChi Key: DENPQNAWGQXKCU-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CS1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)