accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)HN

CAS RN 5817-26-5

IUPAC Name: ethyl (2S)-pyrrolidine-2-carboxylate
Synonyms: ethyl (2S)-pyrrolidine-2-carboxylate
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: QPNJHVDIRZNKOX-LURJTMIESA-N
SMILES: CCOC(=O)[C@@H]1CCCN1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results