accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNOHO

CAS RN 5818-06-4

IUPAC Name: 3-hydroxybenzohydrazide
Synonyms: benzoylhydrazine, M-hydroxy
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: OACGSLLKFCMXSX-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)