accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5819-40-9

CH3ONH2NBr

CAS RN 5819-40-9

IUPAC Name: 4-bromo-5-methyl-1,2-oxazol-3-amine
Synonyms: 4-bromo-5-methyl-1,2-oxazol-3-amine
Molecular Weight (g/mol): 177
Molecular Formula: C4H5BrN2O
InChi Key: JEZOZNWEHSNXPQ-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(N)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results