accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58196-33-1

IUPAC Name: 1,2,3,4-tetrahydroquinolin-7-ol
Synonyms: 1,2,3,4-tetrahydroquinolin-7-ol
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: HJJRGZMJZDSMDB-UHFFFAOYSA-N
SMILES: OC1=CC=C2CCCNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results