accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58197-03-8

IUPAC Name: 2-(thiophen-2-yl)ethanethioamide
Synonyms: 2-(thiophen-2-yl)ethanethioamide
Molecular Weight (g/mol): 157.25
Molecular Formula: C6H7NS2
InChi Key: LUKDARXVJHRFMQ-UHFFFAOYSA-N
SMILES: NC(=S)CC1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results