accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNCH3

CAS RN 582-60-5

IUPAC Name: 5,6-dimethyl-1H-1,3-benzodiazole
Synonyms: 5,6-dimethylbenzimidazole|dimethylbenzimidazole|dimesol|dimedazol|dimezol|dimedazole
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: LJUQGASMPRMWIW-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C2N=CNC2=C1
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)