accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5821-51-2

IUPAC Name: hexacyclo[11.5.2.0⁴,¹⁷.0⁷,¹⁶.0¹⁰,¹⁵.0¹⁴,¹⁸]icosa-1,3,5,7,9,11,13,15,17,19-decaene
Synonyms: corannulene
Molecular Weight (g/mol): 250.3
Molecular Formula: C20H10
InChi Key: VXRUJZQPKRBJKH-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C3C=CC4=C5C6=C(C2=C35)C1=CC=C6C=C4
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)