accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58236-70-7

IUPAC Name: 5-bromo-3-chloro-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-3-chloro-1H-pyridin-2-one
Molecular Weight (g/mol): 208.44
Molecular Formula: C5H3BrClNO
InChi Key: IPWYUROVLMVBGD-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CNC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results