accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(Z)

CAS RN 58296-81-4

IUPAC Name: (8Z)-undec-8-enal
Synonyms: 8-undecenal
Molecular Weight (g/mol): 168.28
Molecular Formula: C11H20O
InChi Key: DWQXOILNGUCNSH-ARJAWSKDSA-N
SMILES: CC\C=C/CCCCCCC=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results