accessibility menu, dialog, popup

UserName

Viewing profile as user:
SHNHN

CAS RN 583-39-1

IUPAC Name: 2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 2-benzimidazolethiol
Molecular Weight (g/mol): 150.2
Molecular Formula: C7H6N2S
InChi Key: YHMYGUUIMTVXNW-UHFFFAOYSA-N
SMILES: S=C1NC2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (2)
  • (1)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (3)
  • (7)
  • (2)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)