accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 583-61-9

IUPAC Name: 2,3-dimethylpyridine
Synonyms: 2,3-dimethylpyridine
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: HPYNZHMRTTWQTB-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1C
Boiling Point 
  • (3)
  • (1)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)