accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOI

CAS RN 58313-23-8

IUPAC Name: ethyl 3-iodobenzoate
Synonyms: ethyl 3-iodobenzoate
Molecular Weight (g/mol): 276.07
Molecular Formula: C9H9IO2
InChi Key: POGCXCWRMMXDAQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC(I)=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results