accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOH

CAS RN 58327-60-9

IUPAC Name: 7-propylquinolin-8-ol
Synonyms: 7-propylquinolin-8-ol
Molecular Weight (g/mol): 187.24
Molecular Formula: C12H13NO
InChi Key: OKPSVYHYNXVYED-UHFFFAOYSA-N
SMILES: CCCC1=CC=C2C=CC=NC2=C1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results