accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 58328-31-7

IUPAC Name: 9-[4'-(9H-carbazol-9-yl)-[1,1'-biphenyl]-4-yl]-9H-carbazole
Synonyms: 9-[4'-(carbazol-9-yl)-[1,1'-biphenyl]-4-yl]carbazole
Molecular Weight (g/mol): 484.6
Molecular Formula: C36H24N2
InChi Key: VFUDMQLBKNMONU-UHFFFAOYSA-N
SMILES: C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (1)
Quantity 
  • (4)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)