accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58328-39-5

IUPAC Name: 5-bromo-2,3-dihydro-1,4-benzodioxine
Synonyms: 5-bromo-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: NGOZRIZXELGVHK-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1OCCO2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results