accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOONOONOONOO

CAS RN 5833-18-1

IUPAC Name: ethyl 2,2-bis(2,4-dinitrophenyl)acetate
Synonyms: ethyl 2,2-bis(2,4-dinitrophenyl)acetate
Molecular Weight (g/mol): 420.29
Molecular Formula: C16H12N4O10
InChi Key: CMVIALRRHVVWIW-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results