accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58336-35-9

IUPAC Name: 3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: 3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Molecular Weight (g/mol): 241.29
Molecular Formula: C15H15NO2
InChi Key: MZSOXGPKUOAXNY-UHFFFAOYSA-N
SMILES: O=C1OC2=C3CCCN4CCCC(C=C2C=C1)=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results