CAS RN 583844-38-6
CAS RN 583844-38-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 1,5-Cyclooctadiene{[Dibenzyl((4S,5S)-5-Methyl-2-Phenyl-4,5-Dihydro-4-Oxazolyl)Methyl]Dicyclohexylphosphinite Kappan:Kappap}Iridium(I) Tetrakis(3,5-Bis(Trifluoromethyl)Phenyl)Borate
Sigma Aldrich 1,5-Cyclooctadiene{[Dibenzyl((4S,5S)-5-Methyl-2-Phenyl-4,5-Dihydro-4-Oxazolyl)Methyl]Dicyclohexylphosphinite Kappan:Kappap}Iridium(I) Tetrakis(3,5-Bis(Trifluoromethyl)Phenyl)Borate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.