accessibility menu, dialog, popup

UserName

Viewing profile as user:
RbRbOOO

CAS RN 584-09-8

IUPAC Name: dirubidium(1+) carbonate
Synonyms: rubidium carbonate
Molecular Weight (g/mol): 230.94
Molecular Formula: CO3Rb2
InChi Key: WPFGFHJALYCVMO-UHFFFAOYSA-L
SMILES: [Rb+].[Rb+].[O-]C([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (5)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (10)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)