accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(S)OOFFF

CAS RN 584-20-3

IUPAC Name: (3S)-3-azaniumyl-4,4,4-trifluorobutanoate
Synonyms: (3S)-3-ammonio-4,4,4-trifluorobutanoate
Molecular Weight (g/mol): 157.09
Molecular Formula: C4H6F3NO2
InChi Key: XRXQOEWWPPJVII-REOHCLBHSA-N
SMILES: [NH3+][C@@H](CC([O-])=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)